Learn More
2-Chlorothiazole, 97%, Thermo Scientific Chemicals
£26.50 - £173.00
Chemical Identifiers
CAS | 3034-52-4 |
---|---|
Molecular Formula | C3H2ClNS |
Molecular Weight (g/mol) | 119.57 |
InChI Key | KLEYVGWAORGTIT-UHFFFAOYSA-N |
Synonym | 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 |
PubChem CID | 76429 |
ChEBI | CHEBI:39187 |
IUPAC Name | 2-chloro-1,3-thiazole |
SMILES | C1=CSC(=N1)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3034-52-4 | |
119.57 | |
2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 | |
CHEBI:39187 | |
C1=CSC(=N1)Cl |
C3H2ClNS | |
KLEYVGWAORGTIT-UHFFFAOYSA-N | |
76429 | |
2-chloro-1,3-thiazole |
Specifications
3034-52-4 | |
145.0°C to 146.0°C | |
Authentic | |
Glass Bottle | |
1.378 | |
KLEYVGWAORGTIT-UHFFFAOYSA-N | |
2-chloro-1,3-thiazole | |
76429 | |
119.57 | |
2-Chlorothiazole |
1.3780g/mL | |
54°C | |
96% min. (GC) | |
C3H2ClNS | |
2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569 | |
C1=CSC(=N1)Cl | |
119.57 | |
CHEBI:39187 | |
97% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 221-228-9
- 2-Chlorothiazole, 97%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.