missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylthiophene, 97%, Thermo Scientific™

£70.42 - £243.00

Chemical Identifiers

CAS 14345-97-2
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.61
MDL Number MFCD00130083
InChI Key KQFADYXPELMVHE-UHFFFAOYSA-N
Synonym 2-chlor-3-methylthiophen, thiophene, 2-chloro-3-methyl, pubchem5164, acmc-209cqk, 2-chloro 3-methyl thiophene, 2-chloro-3-methyl thiophene, ksc174m0h, 2-chloro-3-methylthiophene
PubChem CID 84365
IUPAC Name 2-chloro-3-methylthiophene
SMILES CC1=C(Cl)SC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11317549
View Documents
Thermo Scientific Alfa Aesar
H27957.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11327549
View Documents
Thermo Scientific Alfa Aesar
H27957.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

14345-97-2
132.61
KQFADYXPELMVHE-UHFFFAOYSA-N
84365
CC1=C(Cl)SC=C1
C5H5ClS
MFCD00130083
2-chlor-3-methylthiophen, thiophene, 2-chloro-3-methyl, pubchem5164, acmc-209cqk, 2-chloro 3-methyl thiophene, 2-chloro-3-methyl thiophene, ksc174m0h, 2-chloro-3-methylthiophene
2-chloro-3-methylthiophene

Specifications

14345-97-2
147°C to 149°C
C5H5ClS
MFCD00130083
2-chlor-3-methylthiophen, thiophene, 2-chloro-3-methyl, pubchem5164, acmc-209cqk, 2-chloro 3-methyl thiophene, 2-chloro-3-methyl thiophene, ksc174m0h, 2-chloro-3-methylthiophene
CC1=C(Cl)SC=C1
132.61
132.61
2-Chloro-3-methylthiophene
1.211
46°C (115°F)
1.54
UN1993
KQFADYXPELMVHE-UHFFFAOYSA-N
2-chloro-3-methylthiophene
84365
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapor.
Harmful if inhaled.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 238-296-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications