missing translation for 'onlineSavingsMsg'
Learn More

2-Bromononane, 97%, Thermo Scientific Chemicals

£22.41 - £61.81

Chemical Identifiers

CAS 2216-35-5
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.155
MDL Number MFCD00017865
InChI Key JQEFZTLHNWFZDD-UHFFFAOYSA-N
Synonym sec-nonyl bromide, nonane, 2-bromo, acmc-1cs41
PubChem CID 98219
IUPAC Name 2-bromononane
SMILES CCCCCCCC(C)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361887
View Documents
Thermo Scientific Alfa Aesar
L07387.03
1 g
£22.41
1g
Please to purchase this item. Need a web account? Register with us today!
11371887
View Documents
Thermo Scientific Alfa Aesar
L07387.06
5 g
£61.81
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2216-35-5
207.155
JQEFZTLHNWFZDD-UHFFFAOYSA-N
98219
CCCCCCCC(C)Br
C9H19Br
MFCD00017865
sec-nonyl bromide, nonane, 2-bromo, acmc-1cs41
2-bromononane

Specifications

2216-35-5
208°C to 209°C
1.453
1697559
JQEFZTLHNWFZDD-UHFFFAOYSA-N
2-bromononane
98219
97%
1.081
C9H19Br
MFCD00017865
sec-nonyl bromide, nonane, 2-bromo, acmc-1cs41
CCCCCCCC(C)Br
207.155
207.16
2-Bromononane
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 218-688-8

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.