missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine, 97%, Thermo Scientific™

Product Code. 10376804
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 869901-04-2
Molecular Formula C10H13BrN2
Molecular Weight (g/mol) 241.132
MDL Number MFCD09817465
InChI Key UFXKRINLXMWRES-UHFFFAOYSA-N
Synonym 2-bromomethyl-6-pyrrolidin-1-yl pyridine, 2-bromomethyl-6-pyrrolidin-1-ylpyridine, pyridine,2-bromomethyl-6-1-pyrrolidinyl, 6-bromomethyl-2-pyrrolidinylpyridine, 1-6-bromomethyl pyridin-2-yl pyrrolidine, pyridine, 2-bromomethyl-6-1-pyrrolidinyl
PubChem CID 24229482
IUPAC Name 2-(bromomethyl)-6-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=CC=CC(=N2)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 241.13
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)-6-pyrrolidin-1-ylpyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.