Learn More
2-Bromo-2',4'-difluoroacetophenone, 97%, Thermo Scientific™
£51.64 - £184.00
Chemical Identifiers
CAS | 102429-07-2 |
---|---|
Molecular Formula | C8H5BrF2O |
Molecular Weight (g/mol) | 235.028 |
MDL Number | MFCD00142822 |
InChI Key | CSGDTHXBRAAOHV-UHFFFAOYSA-N |
Synonym | 2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw |
PubChem CID | 2774185 |
IUPAC Name | 2-bromo-1-(2,4-difluorophenyl)ethanone |
SMILES | C1=CC(=C(C=C1F)F)C(=O)CBr |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
102429-07-2 | |
235.028 | |
CSGDTHXBRAAOHV-UHFFFAOYSA-N | |
2774185 | |
C1=CC(=C(C=C1F)F)C(=O)CBr |
C8H5BrF2O | |
MFCD00142822 | |
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw | |
2-bromo-1-(2,4-difluorophenyl)ethanone |
Specifications
102429-07-2 | |
51°C (124°F) | |
MFCD00142822 | |
Air sensitive | |
CSGDTHXBRAAOHV-UHFFFAOYSA-N | |
2-bromo-1-(2,4-difluorophenyl)ethanone | |
2774185 | |
97% |
31°C to 35°C | |
C8H5BrF2O | |
UN2925 | |
2-bromo-2',4'-difluoroacetophenone, 2-bromo-1-2,4-difluorophenyl ethanone, 2,4-difluorophenacyl bromide, 2-bromo-1-2,4-difluorophenyl ethan-1-one, 2-bromo-1-2,4-difluoro-phenyl-ethanone, 2',4'-difluorophenacyl bromide, 2-bromo-2,4-difluoroacetophenone, ethanone, 2-bromo-1-2,4-difluorophenyl, alpha-bromo-2',4'-difluoroacetophenone, acmc-1c4aw | |
C1=CC(=C(C=C1F)F)C(=O)CBr | |
235.028 | |
235.03 | |
2-Bromo-2',4'-difluoroacetophenone |
Safety and Handling
GHS H Statement
H228-H314-H318-H302
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H228-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
Your input is important to us. Please complete this form to provide feedback related to the content on this product.