missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethan-1-one, Thermo Scientific™
£85.10 - £274.00
Chemical Identifiers
CAS | 175205-89-7 |
---|---|
Molecular Formula | C9H4BrCl2F3O |
Molecular Weight (g/mol) | 335.93 |
MDL Number | MFCD00221016 |
InChI Key | MSMAFPRJZLLQNJ-UHFFFAOYSA-N |
Synonym | 2-bromo-2',6'-dichloro-4'-trifluoromethyl-acetophenone, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethanone, ethanone,2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl, 2-bromo-2,6-dichloro-4-trifluoromethyl-acetophenone, 2-bromo-2',6'-dichloro-4'-trifluoromethyl acetophenone, 1-2,6-bis chloranyl-4-trifluoromethyl phenyl-2-bromanyl-ethanone |
PubChem CID | 2781609 |
IUPAC Name | 2-bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethanone |
SMILES | FC(F)(F)C1=CC(Cl)=C(C(=O)CBr)C(Cl)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10059471
|
Thermo Scientific Maybridge
SEW04510DA |
1 g | Amber Glass Bottle |
£85.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10143234
|
Thermo Scientific Maybridge
SEW04510DE |
5 g | Amber Glass Bottle |
£274.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175205-89-7 | |
335.93 | |
MSMAFPRJZLLQNJ-UHFFFAOYSA-N | |
2781609 | |
FC(F)(F)C1=CC(Cl)=C(C(=O)CBr)C(Cl)=C1 |
C9H4BrCl2F3O | |
MFCD00221016 | |
2-bromo-2',6'-dichloro-4'-trifluoromethyl-acetophenone, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethanone, ethanone,2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl, 2-bromo-2,6-dichloro-4-trifluoromethyl-acetophenone, 2-bromo-2',6'-dichloro-4'-trifluoromethyl acetophenone, 1-2,6-bis chloranyl-4-trifluoromethyl phenyl-2-bromanyl-ethanone | |
2-bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethanone |
Specifications
175205-89-7 | |
C9H4BrCl2F3O | |
2-bromo-2',6'-dichloro-4'-trifluoromethyl-acetophenone, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethan-1-one, 2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl ethanone, ethanone,2-bromo-1-2,6-dichloro-4-trifluoromethyl phenyl, 2-bromo-2,6-dichloro-4-trifluoromethyl-acetophenone, 2-bromo-2',6'-dichloro-4'-trifluoromethyl acetophenone, 1-2,6-bis chloranyl-4-trifluoromethyl phenyl-2-bromanyl-ethanone | |
FC(F)(F)C1=CC(Cl)=C(C(=O)CBr)C(Cl)=C1 | |
335.93 | |
335.93 | |
2-Bromo-1-[2, 6-dichloro-4-(trifluoromethyl)phenyl]ethan-1-one |
95% | |
MFCD00221016 | |
MSMAFPRJZLLQNJ-UHFFFAOYSA-N | |
2-bromo-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]ethanone | |
2781609 | |
95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title