missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)2C6H3C(O)CH2Br |
Quantity | 10 g |
Formula Weight | 227.1 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction