missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Benzoylpyridine, 99%

£28.20 - £102.00

Chemical Identifiers

CAS 91-02-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006300
InChI Key GCSHUYKULREZSJ-UHFFFAOYSA-N
Synonym 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone
PubChem CID 7038
IUPAC Name phenyl(pyridin-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=N2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11489832
SDS View Documents
Alfa Aesar™
A10857.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11499832
SDS View Documents
Alfa Aesar™
A10857.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Applications
The Schiff bases(2-Benzoylpyridine ) and copper(II) complexes display moderate antifungal activities. 2-Benzoylpyridine is an aromatic substituent which has selective antiproliferative effects, with the majority having greater antineoplastic activity. Reaction of 2-benzoylpyridine N(4)-phenyl thiosemicarbazone (HL) with a variety of copper salts yielded a series of copper(II) complexes of the type [CuLX]. Complexes of 2-benzoylpyridine thiosemicarbazone (H2Bz4DH) as well as of its N(4)-methyl (H2Bz4Me) and N(4)-phenyl (H2Bz4Ph) analogues with iron(II), nickel(II) and zinc(II) were characterized by a variety of spectroscopic techniques.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

91-02-1
183.21
GCSHUYKULREZSJ-UHFFFAOYSA-N
7038
C1=CC=C(C=C1)C(=O)C2=CC=CC=N2
C12H9NO
MFCD00006300
2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone
phenyl(pyridin-2-yl)methanone

Specifications

91-02-1
313°C
C12H9NO
120283
GCSHUYKULREZSJ-UHFFFAOYSA-N
phenyl(pyridin-2-yl)methanone
7038
99%
41°C to 45°C
167°C (332°F)
MFCD00006300
2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=N2
183.21
183.21
2-Benzoylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'rtecsNumber' : OB6400000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.