missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine-5-boronic acid, 97%, Thermo Scientific™

Product Code. 10708635
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 936250-22-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C4H6BN3O2
MDL Number MFCD07375147
Quantity 1 g
Synonym 2-aminopyrimidine-5-boronic acid, 2-aminopyrimidin-5-yl boronic acid, 2-amino-pyrimidine-5-boronic acid, 2-amino-5-pyrimidineboronic acid, 2-aminopyrimidin-5-yl-5-boronic acid, 2-amino-5-boronopyrimidine, acmc-209rmr, 2-aminopyrimidine 5-boronic acid, 2-amino-5-pyrimidinylboronic acid, 2-amino-pyrimidin-5-ylboronic acid
InChI Key CGHYQZASLKERLV-UHFFFAOYSA-N
SMILES NC1=NC=C(C=N1)B(O)O
IUPAC Name (2-aminopyrimidin-5-yl)boronic acid
Molecular Weight (g/mol) 138.92
PubChem CID 17750211
Formula Weight 138.92
Percent Purity 97%
Chemical Name or Material 2-Aminopyrimidine-5-boronic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'