Learn More
2-Amino-6-picoline, 98%, Thermo Scientific Chemicals
£20.90 - £423.94
Chemical Identifiers
CAS | 1824-81-3 |
---|---|
Molecular Formula | C6H8N2 |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00006331 |
InChI Key | QUXLCYFNVNNRBE-UHFFFAOYSA-N |
Synonym | 2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl |
PubChem CID | 15765 |
IUPAC Name | 6-methylpyridin-2-amine |
SMILES | CC1=CC=CC(N)=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10409590
|
Thermo Scientific Acros
104400050 |
5 g | Glass bottle |
£20.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10754401
|
Thermo Scientific Acros
104401000 |
100 g | Glass bottle |
£27.74
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10754591
|
Thermo Scientific Acros
104405000 |
500 g | Glass bottle |
£103.52
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626914
|
Thermo Scientific Acros
104400025 |
2.5 kg | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1824-81-3 | |
108.14 | |
QUXLCYFNVNNRBE-UHFFFAOYSA-N | |
15765 | |
CC1=CC=CC(N)=N1 |
C6H8N2 | |
MFCD00006331 | |
2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl | |
6-methylpyridin-2-amine |
Specifications
1824-81-3 | |
100.0 | |
Yellow | |
103°C | |
97.5% min. (GC) | |
MFCD00006331 | |
2-amino-6-methylpyridine, 2-amino-6-picoline, 6-amino-2-picoline, 2-pyridinamine, 6-methyl, 6-methyl-2-pyridinamine, 6-methyl-2-pyridylamine, 6-methyl-2-aminopyridine, 2-picoline, 6-amino, 6-amino-2-methylpyridine, pyridine, 2-amino-6-methyl | |
QUXLCYFNVNNRBE-UHFFFAOYSA-N | |
6-methylpyridin-2-amine | |
15765 | |
98% | |
2-Amino-6-picoline, 98% |
97.5 | |
40°C to 45°C | |
208°C to 209°C | |
Authentic | |
C6H8N2 | |
22, V,9, 210 | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohols and chlorinated solvents,sparingly soluble in toluene | |
CC1=CC=CC(N)=N1 | |
108.14 | |
108.14 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 217-360-1
missing translation for 'rtecsNumber' : US1885000
missing translation for 'tsca' : TSCA
- 2-Amino-6-picoline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only