Learn More
(+/-)-2-Amino-1-propanol, 98%, Thermo Scientific Chemicals
£32.70 - £102.00
Chemical Identifiers
CAS | 6168-72-5 |
---|---|
Molecular Formula | C3H9NO |
Molecular Weight (g/mol) | 75.11 |
MDL Number | MFCD00008085 |
InChI Key | BKMMTJMQCTUHRP-UHFFFAOYNA-N |
Synonym | dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine |
PubChem CID | 5126 |
IUPAC Name | 2-aminopropan-1-ol |
SMILES | CC(N)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322236
|
Thermo Scientific Alfa Aesar
L03608.06 |
5 g |
£32.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332236
|
Thermo Scientific Alfa Aesar
L03608.14 |
25 g |
£102.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.
Solubility
Fully miscible in water.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
Chemical Identifiers
6168-72-5 | |
75.11 | |
BKMMTJMQCTUHRP-UHFFFAOYNA-N | |
5126 | |
CC(N)CO |
C3H9NO | |
MFCD00008085 | |
dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine | |
2-aminopropan-1-ol |
Specifications
6168-72-5 | |
0.965 | |
83°C (181°F) | |
1.4495 | |
UN2735 | |
Air sensitive; Hygroscopic | |
Fully miscible in water. | |
CC(N)CO | |
75.11 | |
75.11 | |
(+/-)-2-Amino-1-propanol |
8°C | |
174°C to 176°C | |
C3H9NO | |
MFCD00008085 | |
1209234 | |
dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine | |
BKMMTJMQCTUHRP-UHFFFAOYNA-N | |
2-aminopropan-1-ol | |
5126 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 228-207-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only