missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-methoxybenzonitrile, 97%, Thermo Scientific™

£94.83 - £314.00

Chemical Identifiers

CAS 886498-35-7
Molecular Formula C8H5F2NO
Molecular Weight (g/mol) 169.131
MDL Number MFCD04115902
InChI Key BXVCAGRBLOMHBH-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-methoxy-benzonitrile, 2,6-difluoro-3-methoxybenzenecarbonitrile
PubChem CID 3791832
IUPAC Name 2,6-difluoro-3-methoxybenzonitrile
SMILES COC1=C(C(=C(C=C1)F)C#N)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11386859
SDS View Documents
Thermo Scientific Alfa Aesar
H26775.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11396859
SDS View Documents
Thermo Scientific Alfa Aesar
H26775.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

886498-35-7
169.131
BXVCAGRBLOMHBH-UHFFFAOYSA-N
3791832
COC1=C(C(=C(C=C1)F)C#N)F
C8H5F2NO
MFCD04115902
2,6-difluoro-3-methoxy-benzonitrile, 2,6-difluoro-3-methoxybenzenecarbonitrile
2,6-difluoro-3-methoxybenzonitrile

Specifications

886498-35-7
C8H5F2NO
UN3439
BXVCAGRBLOMHBH-UHFFFAOYSA-N
2,6-difluoro-3-methoxybenzonitrile
3791832
97%
71°C to 73°C
MFCD04115902
2,6-difluoro-3-methoxy-benzonitrile, 2,6-difluoro-3-methoxybenzenecarbonitrile
COC1=C(C(=C(C=C1)F)C#N)F
169.131
169.13
2,6-Difluoro-3-methoxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.