Learn More
Fisher Chemical 2.5 LT ETHANOL,100%,CERTIFIED HPLC 2.5 LITRES
Brand: Fisher Chemical E/0665/17
200.49 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
2.5 LT ETHANOL,100%,CERTIFIED HPLC 2.5 LITRES
Description

Safety and Handling
- Ethanol, absolute
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Ethanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2531988 | Certificate of Analysis | 25/03/2025 | E/0665DF/17Z,E/0665DF/15X,E/0665DF/17,E/0665DF/27Z,E/0665DF/17X,E/0665DF/15,E/0665DF/17CN,E/0665/17,E/0665/15 |
BS25/009 | Certificate of Analysis | 19/03/2025 | E/0665DF/17Z,E/0665DF/15X,E/0665DF/17,E/0665DF/27Z,E/0665DF/17X,E/0665DF/15,E/0665DF/17CN,E/0665/17,E/0665/15 |
2528690 | Certificate of Analysis | 26/02/2025 | E/0665DF/17Z,E/0665DF/15X,E/0665DF/17,E/0665DF/27Z,E/0665DF/17X,E/0665DF/15,E/0665DF/17CN,E/0665/17,E/0665/15,E/0665DF/P17 |
2526667 | Certificate of Analysis | 11/02/2025 | E/0665DF/17Z,E/0665DF/17,E/0665DF/15X,E/0665DF/27Z,E/0665DF/17X,E/0665DF/15,E/0665DF/17CN,E/0665/17,E/0665/15,E/0665DF/P17 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.