missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2′,5′-Dihydroxyacetophenone, ≥99.5%, Honeywell Fluka™
Matrix substance for MALDI-MS,≥99.5%
£19.67 - £54.33
Chemical Identifiers
CAS | 490-78-8 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00002343 |
InChI Key | WLDWSGZHNBANIO-UHFFFAOYSA-N |
Synonym | 2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy |
PubChem CID | 10279 |
IUPAC Name | 1-(2,5-dihydroxyphenyl)ethanone |
SMILES | CC(=O)C1=C(C=CC(=C1)O)O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
490-78-8 | |
152.149 | |
WLDWSGZHNBANIO-UHFFFAOYSA-N | |
10279 | |
CC(=O)C1=C(C=CC(=C1)O)O |
C8H8O3 | |
MFCD00002343 | |
2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy | |
1-(2,5-dihydroxyphenyl)ethanone |
Specifications
490-78-8 | |
Yellow | |
(HO)2C6H3COCH3 | |
NONH for all modes of transport | |
2',5'-dihydroxyacetophenone, 1-2,5-dihydroxyphenyl ethanone, 2-acetylhydroquinone, 2,5-dihydroxyacetophenone, quinacetophenone, acetylhydroquinone, ethanone, 1-2,5-dihydroxyphenyl, 1-2,5-dihydroxyphenyl ethan-1-one, acetylquinol, acetophenone, 2',5'-dihydroxy | |
CC(=O)C1=C(C=CC(=C1)O)O | |
152.149 | |
152.15g/mol | |
2′,5′-Dihydroxyacetophenone |
204°C to 207°C | |
C8H8O3 | |
MFCD00002343 | |
637903 | |
WLDWSGZHNBANIO-UHFFFAOYSA-N | |
1-(2,5-dihydroxyphenyl)ethanone | |
10279 | |
≥99.5% |
Safety and Handling
missing translation for 'rtecsNumber' : 207-716-4
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title