Learn More
2,5-Dichloropyrimidine, 95%, Thermo Scientific Chemicals
£31.66 - £354.77
Chemical Identifiers
CAS | 22536-67-0 |
---|---|
Molecular Formula | C4H2Cl2N2 |
Molecular Weight (g/mol) | 148.974 |
MDL Number | MFCD00673131 |
InChI Key | CEJAHXLRNZJPQH-UHFFFAOYSA-N |
Synonym | 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci |
PubChem CID | 590641 |
IUPAC Name | 2,5-dichloropyrimidine |
SMILES | C1=C(C=NC(=N1)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15431099
|
Thermo Scientific Alfa Aesar
H64066.06 |
5 g |
£31.66
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15441099
|
Thermo Scientific Alfa Aesar
H64066.14 |
25 g |
£94.77
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451099
|
Thermo Scientific Alfa Aesar
H64066.22 |
100 g |
£354.77
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
22536-67-0 | |
148.974 | |
CEJAHXLRNZJPQH-UHFFFAOYSA-N | |
590641 | |
C1=C(C=NC(=N1)Cl)Cl |
C4H2Cl2N2 | |
MFCD00673131 | |
2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci | |
2,5-dichloropyrimidine |
Specifications
22536-67-0 | |
White | |
C4H2Cl2N2 | |
UN3077 | |
2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci | |
CEJAHXLRNZJPQH-UHFFFAOYSA-N | |
2,5-dichloropyrimidine | |
590641 | |
95% | |
2,5-Dichloropyrimidine |
48°C to 55°C | |
107°C (225°F) | |
MFCD00673131 | |
110912 | |
Slightly soluble in water. | |
C1=C(C=NC(=N1)Cl)Cl | |
148.974 | |
148.97 | |
Powder |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.