missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine-4-carboxylic acid, 95%, Thermo Scientific™

Product Code. 15484598
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88912-26-9
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00466640
InChI Key GFOVTTQVBDEYPP-UHFFFAOYSA-N
Synonym 2,5-dichloroisonicotinic acid, 2,5-dichloro-isonicotinic acid, 4-carboxy-2,5-dichloropyridine, 4-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-4-pyridinecarboxylic acid, pubchem13559, acmc-209qxv, ksc447s3n, 2,5-dichloro isonicotinic acid, gfovttqvbdeypp-uhfffaoysa
PubChem CID 2736087
IUPAC Name 2,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=CN=C1Cl)Cl)C(=O)O

Specifications

Melting Point 235°C to 240°C
Quantity 250 mg
Formula Weight 192
Percent Purity 95%
Chemical Name or Material 2,5-Dichloropyridine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'