missing translation for 'onlineSavingsMsg'
Learn More

2,5-dichlorophenyl isothiocyanate, Thermo Scientific™

£16.10 - £44.90

Chemical Identifiers

CAS 3386-42-3
Molecular Formula C7H3Cl2NS
Molecular Weight (g/mol) 204.068
InChI Key JHTPBGFVWWSHDL-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl isothiocyanate, 2,5-dichlorophenylisothiocyanate, acmc-1cjox, 2,5-dichlorobenzenisothiocyanate, #, benzene,1,4-dichloro-2-isothiocyanato, benzene, 1,4-dichloro-2-isothiocyanato, 1,4-bis chloranyl-2-isothiocyanato-benzene
PubChem CID 137915
IUPAC Name 1,4-dichloro-2-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1Cl)N=C=S)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10026684
View Documents
Thermo Scientific Maybridge
SB01698DE
5 g
£16.10
5g
Please to purchase this item. Need a web account? Register with us today!
10557363
View Documents
Thermo Scientific Maybridge
SB01698EA
10 g
£23.00
10g
Please to purchase this item. Need a web account? Register with us today!
10515004
View Documents
Thermo Scientific Maybridge
SB01698EB
25 g
£44.90
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

3386-42-3
204.068
2,5-dichlorophenyl isothiocyanate, 2,5-dichlorophenylisothiocyanate, acmc-1cjox, 2,5-dichlorobenzenisothiocyanate, #, benzene,1,4-dichloro-2-isothiocyanato, benzene, 1,4-dichloro-2-isothiocyanato, 1,4-bis chloranyl-2-isothiocyanato-benzene
1,4-dichloro-2-isothiocyanatobenzene
C7H3Cl2NS
JHTPBGFVWWSHDL-UHFFFAOYSA-N
137915
C1=CC(=C(C=C1Cl)N=C=S)Cl

Specifications

3386-42-3
Amber glass bottle
2,5-dichlorophenyl isothiocyanate, 2,5-dichlorophenylisothiocyanate, acmc-1cjox, 2,5-dichlorobenzenisothiocyanate, #, benzene,1,4-dichloro-2-isothiocyanato, benzene, 1,4-dichloro-2-isothiocyanato, 1,4-bis chloranyl-2-isothiocyanato-benzene
C1=CC(=C(C=C1Cl)N=C=S)Cl
204.068
204.08
2, 5-Dichlorophenyl isothiocyanate
97+%
C7H3Cl2NS
JHTPBGFVWWSHDL-UHFFFAOYSA-N
1,4-dichloro-2-isothiocyanatobenzene
137915
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications