missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-n-butylthiophene, 96%, Thermo Scientific™

Product Code. 15451718
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116971-10-9
Molecular Formula C8H10Br2S
Molecular Weight (g/mol) 298.04
MDL Number MFCD00672012
InChI Key YRSQCQUZWSQKKU-UHFFFAOYSA-N
PubChem CID 600361
IUPAC Name 2,5-dibromo-3-butylthiophene
SMILES CCCCC1=C(Br)SC(Br)=C1

Specifications

Density 1.67
Boiling Point 270°C to 281°C
Flash Point 110°C (230°F)
Refractive Index 1.574
Quantity 1 g
Synonym thiophene, 2,5-dibromo-3-butyl, acmc-2099tk, ksc491i5n, 3-butyl-2,5-dibromothiophene, 2,5-dibromo-3-butyl-thiophene, 2,5-dibromo-3-n-butylthiophene, thiophene,2,5-dibromo-3-butyl, 2,5-bis bromanyl-3-butyl-thiophene, 2,5-dibromo-3-butylthiophene
Formula Weight 298.04
Percent Purity 96%
Chemical Name or Material 2,5-Dibromo-3-n-butylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'