missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific Chemicals

Product Code. 15402598
Click to view available options
Quantity:
10 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer's disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for preparation of heteroarylpiperazine derivatives for use in treatment of Alzheimer’s disease.

Solubility
Slightly soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19646-07-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179.00
MDL Number MFCD02685670
InChI Key ZTHHRSBDBPCCMZ-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-methoxy-pyrimidine, 2,4-dichloro-5-methoxy pyrimidine, pyrimidine, 2,4-dichloro-5-methoxy, 2,4-dichloropyrimidin-5-yl methyl ether, 5-methoxy-2,4-dichloro pyrimidine, pubchem7076, ksc490s5l, 2,4 dichloro-5-methoxypyrimidine, 2,4-dichloro-5-methoxy-pyrimidin, 2,6-dichloro-5-methoxy pyrimidine
PubChem CID 601401
IUPAC Name 2,4-dichloro-5-methoxypyrimidine
SMILES COC1=CN=C(Cl)N=C1Cl

Specifications

Melting Point 66°C to 70°C
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 179
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-methoxypyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.