missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trinitrotoluene, SPEX CertiPrep™
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
TNT (Solution) | 118-96-7 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
CAS | 118-96-7, 67-56-1 |
CAS Min % | 0.1, 99.9 |
CAS Max % | 0.1, 99.9 |
Molecular Formula | C7H5N3O6 |
InChI Key | SPSSULHKWOKEEL-UHFFFAOYSA-N |
SMILES | CC1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-] |
IUPAC Name | 2-methyl-1,3,5-trinitrobenzene |
Molecular Weight (g/mol) | 227.132 |
PubChem CID | 8376 |
ChEBI | CHEBI:46053 |
Show More |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : methanol, acetonitrile
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'