Learn More
2,3-Dibromothiophene, 98+%, Thermo Scientific Chemicals
£16.20 - £170.00
Chemical Identifiers
CAS | 3140-93-0 |
---|---|
Molecular Formula | C4H2Br2S |
Molecular Weight (g/mol) | 241.928 |
MDL Number | MFCD00005418 |
InChI Key | ATRJNSFQBYKFSM-UHFFFAOYSA-N |
Synonym | thiophene, 2,3-dibromo, 2,3-dibromo thiophene, dibromothiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, 2 3-dibromothiophene, pubchem5511, thiophene,3-dibromo, acmc-1ckid, ksc491i5b |
PubChem CID | 76590 |
IUPAC Name | 2,3-dibromothiophene |
SMILES | C1=CSC(=C1Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11409893
|
Thermo Scientific Alfa Aesar
A12998.03 |
1 g |
£16.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419893
|
Thermo Scientific Alfa Aesar
A12998.06 |
5 g |
£52.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429893
|
Thermo Scientific Alfa Aesar
A12998.14 |
25 g |
£170.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3140-93-0 | |
241.928 | |
ATRJNSFQBYKFSM-UHFFFAOYSA-N | |
76590 | |
C1=CSC(=C1Br)Br |
C4H2Br2S | |
MFCD00005418 | |
thiophene, 2,3-dibromo, 2,3-dibromo thiophene, dibromothiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, 2 3-dibromothiophene, pubchem5511, thiophene,3-dibromo, acmc-1ckid, ksc491i5b | |
2,3-dibromothiophene |
Specifications
3140-93-0 | |
215°C to 216°C | |
C4H2Br2S | |
MFCD00005418 | |
Light sensitive | |
ATRJNSFQBYKFSM-UHFFFAOYSA-N | |
2,3-dibromothiophene | |
76590 | |
≥98% |
2.135 | |
>100°C (212°F) | |
1.631 | |
107512 | |
thiophene, 2,3-dibromo, 2,3-dibromo thiophene, dibromothiophene, 2,3-dibromo-thiophene, 2,3-dibromthiophen, 2 3-dibromothiophene, pubchem5511, thiophene,3-dibromo, acmc-1ckid, ksc491i5b | |
C1=CSC(=C1Br)Br | |
241.928 | |
241.94 | |
2,3-Dibromothiophene |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 221-542-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only