missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%, Thermo Scientific Chemicals

£26.93 - £81.00

Chemical Identifiers

CAS 22195-47-7
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD01321384
InChI Key HXOYWCSTHVTLOW-UHFFFAOYNA-N
Synonym 2,2-dimethyl-1,3-dioxolane-4-methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-methylamine, 1-2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methylamine, 2,2-dimethyl-1,3-dioxolan-4-methanamine, 1,3-dioxolane-4-methanamine, 2,2-dimethyl, 2,2-dimethyl-4-aminomethyl-1,3-dioxolane, 2.2-dimethyl-1,3-dioxolane-4-methanamine
PubChem CID 4122349
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
SMILES CC1(C)OCC(CN)O1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10624755
View Documents
Thermo Scientific Acros
397410010
1 g Glass bottle
£26.93
1g
Please to purchase this item. Need a web account? Register with us today!
10401484
View Documents
Thermo Scientific Acros
397410050
5 g Glass bottle
£81.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22195-47-7
131.18
HXOYWCSTHVTLOW-UHFFFAOYNA-N
4122349
CC1(C)OCC(CN)O1
C6H13NO2
MFCD01321384
2,2-dimethyl-1,3-dioxolane-4-methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-methylamine, 1-2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methylamine, 2,2-dimethyl-1,3-dioxolan-4-methanamine, 1,3-dioxolane-4-methanamine, 2,2-dimethyl, 2,2-dimethyl-4-aminomethyl-1,3-dioxolane, 2.2-dimethyl-1,3-dioxolane-4-methanamine
(2,2-dimethyl-1,3-dioxolan-4-yl)methanamine

Specifications

22195-47-7
100.0
1.0120g/mL
68°C
96% min. (GC)
1.4370 to 1.4390 (20°C, 589nm)
1.012
HXOYWCSTHVTLOW-UHFFFAOYNA-N
(2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
4122349
97%
2, 2-Dimethyl-1, 3-dioxolane-4-methanamine
96.0
Colorless to Yellow
164°C to 165°C
Authentic
C6H13NO2
MFCD01321384
2,2-dimethyl-1,3-dioxolane-4-methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-methylamine, 1-2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methylamine, 2,2-dimethyl-1,3-dioxolan-4-methanamine, 1,3-dioxolane-4-methanamine, 2,2-dimethyl, 2,2-dimethyl-4-aminomethyl-1,3-dioxolane, 2.2-dimethyl-1,3-dioxolane-4-methanamine
CC1(C)OCC(CN)O1
131.18
131.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

Product Identifier
  • 2,2-Dimethyl-1,3-dioxolane-4-methanamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only