Learn More
(+/-)-2,2'-Dibromo-1,1'-binaphthyl, 96%, Thermo Scientific Chemicals
£61.80 - £216.00
Chemical Identifiers
CAS | 74866-28-7 |
---|---|
Molecular Formula | C20H12Br2 |
Molecular Weight (g/mol) | 412.124 |
MDL Number | MFCD00188005 |
InChI Key | IJUDEFZBMMRSNM-UHFFFAOYSA-N |
Synonym | 2,2'-dibromo-1,1'-binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, racemic-2,2'-dibromo-1,1'-binaphthyl, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, s---2,2-dibromo-1,1'-binaphthyl, 2,2'-dibromo 1,1' binaphthyl, 1,1'-bi 2-bromonaphthalene |
PubChem CID | 619909 |
IUPAC Name | 2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene |
SMILES | C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441620
|
Thermo Scientific Alfa Aesar
H26987.03 |
1 g |
£61.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451620
|
Thermo Scientific Alfa Aesar
H26987.06 |
5 g |
£216.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The synthesis of this class of compounds relies on the lithiation of racemic 2,2'-dibromo-1,1'-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe synthesis of this class of compounds relies on the lithiation of racemic 2,2′-dibromo-1,1′-binaphthyl with t-BuLi followed by addition of Ph 2 PCl.
Solubility
Sparingly Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
74866-28-7 | |
412.124 | |
IJUDEFZBMMRSNM-UHFFFAOYSA-N | |
619909 | |
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br |
C20H12Br2 | |
MFCD00188005 | |
2,2'-dibromo-1,1'-binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, racemic-2,2'-dibromo-1,1'-binaphthyl, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, s---2,2-dibromo-1,1'-binaphthyl, 2,2'-dibromo 1,1' binaphthyl, 1,1'-bi 2-bromonaphthalene | |
2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene |
Specifications
74866-28-7 | |
C20H12Br2 | |
2,2'-dibromo-1,1'-binaphthyl, 2,2'-dibromo-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-dibromo, racemic-2,2'-dibromo-1,1'-binaphthyl, 1r-2,2'-dibromo-1,1'-binaphthalene, 1s-2,2'-dibromo-1,1'-binaphthalene, 2-bromo-1-2-bromonaphthalen-1-yl naphthalene, s---2,2-dibromo-1,1'-binaphthyl, 2,2'-dibromo 1,1' binaphthyl, 1,1'-bi 2-bromonaphthalene | |
IJUDEFZBMMRSNM-UHFFFAOYSA-N | |
2-bromo-1-(2-bromonaphthalen-1-yl)naphthalene | |
619909 | |
96% |
178°C to 183°C | |
MFCD00188005 | |
Sparingly Soluble in water. | |
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)Br)Br | |
412.124 | |
412.23 | |
(+/-)-2,2'-Dibromo-1,1'-binaphthyl |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.