missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-chlorophenoxy)ethylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Quantity | 1 g |
Packaging | Amber Glass Bottle |
Assay Percent Range | 97% |
Linear Formula | ClC6H4O(CH2)2NH2 |
Synonym | 2-2-chlorophenoxy ethylamine, 2-2-chlorophenoxy ethanamine, 2-2-chlorophenoxy-1-ethanamine, ethanamine, 2-2-chlorophenoxy, 2-2-chlorophenoxy ethyl amine, 1-2-aminoethoxy-2-chlorobenzene, acmc-20aoqz, ethanamine,2-2-chlorophenoxy, 2-2-chlorophenoxy ethan-1-amine |
Formula Weight | 171.63 |
Percent Purity | 97% |
Chemical Name or Material | 2-(2-Chlorophenoxy)ethylamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction