missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Chloroethoxy)ethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-7020947
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
628-89-7
C4H9ClO2
124.56
MFCD00002870
LECMBPWEOVZHKN-UHFFFAOYSA-N
2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210
12361
2-(2-chloroethoxy)ethanol
OCCOCCCl
This item is not returnable. View return policy
628-89-7
C4H9ClO2
124.56
MFCD00002870
LECMBPWEOVZHKN-UHFFFAOYSA-N
2-2-chloroethoxy ethanol, ethanol, 2-2-chloroethoxy, diglycol chlorohydrin, diglycol chlorhydrin, 2-chloroethyl 2-hydroxyethyl ether, monochloropolyoxyethylene, 2-2-chloroethoxy ethan-1-ol, unii-r2up818l4i, diethylene glycol monochlorohydrin, polyethylene glycol chloride 210
12361
2-(2-chloroethoxy)ethanol
OCCOCCCl

2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Chloroethoxy)ethanol is employed in the preparation of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine and 2-(2-azidoethoxy)ethanol.

Solubility
Miscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Density 1.18
Boiling Point 185°C
Flash Point 90°C (194°F)
Linear Formula ClCH2CH2OCH2CH2OH
Refractive Index 1.4535
Quantity 50 g
Beilstein 506015
Solubility Information Miscible with water.
Formula Weight 124.57
Percent Purity 99%
Chemical Name or Material 2-(2-Chloroethoxy)ethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'