Learn More
2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals
£48.64 - £767.82
Chemical Identifiers
CAS | 6976-37-0 |
---|---|
Molecular Formula | C8H19NO5 |
Molecular Weight (g/mol) | 209.24 |
MDL Number | MFCD00002853 |
InChI Key | OWMVSZAMULFTJU-UHFFFAOYSA-N |
Synonym | bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane |
PubChem CID | 81462 |
ChEBI | CHEBI:41250 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(CO)N(CCO)C(CO)(CO)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10134130
|
Thermo Scientific Acros
163821000 |
100 g |
£48.64
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10434955
|
Thermo Scientific Acros
163825000 |
500 g |
£163.82
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10791935
|
Thermo Scientific Acros
163820025 |
2.5 kg |
£767.82
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6976-37-0 | |
209.24 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
81462 | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |
C8H19NO5 | |
MFCD00002853 | |
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane | |
CHEBI:41250 | |
C(CO)N(CCO)C(CO)(CO)CO |
Specifications
6976-37-0 | |
97.5 | |
103.0°C to 104.0°C | |
0.2% max. (80°C, 5 hrs) | |
98% | |
C8H19NO5 | |
MFCD00002853 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.24 | |
CHEBI:41250 | |
98% | |
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 98% |
(0.1M in H2O) | |
100.0 | |
White to Brown | |
Authentic | |
Plastic bottle | |
(HOCH2CH2)2NC(CH2OH)3 | |
bis-tris, bistris, 2,2-bis hydroxymethyl-2,2',2-nitrilotriethanol, bis 2-hydroxyethyl aminotris hydroxymethyl methane, bis-tris buffer, 1,3-propanediol, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl, unii-q1xc3631cp, bis 2-hydroxyethyl amino-tris hydroxymethyl methane, 2-bis 2-hydroxyethyl amino-2-hydroxymethyl propane-1,3-diol, bis 2-hydroxyethyl iminotris hydroxymethyl methane | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
209.24 | |
Crystalline Powder or Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 230-237-7
missing translation for 'tsca' : TSCA
- 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.