Learn More
2,2',4'-Trichloroacetophenone, 97%, Thermo Scientific Chemicals
£17.59 - £203.01
Chemical Identifiers
CAS | 4252-78-2 |
---|---|
Molecular Formula | C8H5Cl3O |
Molecular Weight (g/mol) | 223.477 |
MDL Number | MFCD00000934 |
InChI Key | VYWPPRLJNVHPEU-UHFFFAOYSA-N |
Synonym | 2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one |
PubChem CID | 20250 |
IUPAC Name | 2-chloro-1-(2,4-dichlorophenyl)ethanone |
SMILES | C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11469792
|
Thermo Scientific Alfa Aesar
A10835.14 |
25 g |
£17.59
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479792
|
Thermo Scientific Alfa Aesar
A10835.22 |
100 g |
£52.46
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489792
|
Thermo Scientific Alfa Aesar
A10835.36 |
500 g |
£203.01
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4252-78-2 | |
223.477 | |
VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
20250 | |
C1=CC(=C(C=C1Cl)Cl)C(=O)CCl |
C8H5Cl3O | |
MFCD00000934 | |
2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
2-chloro-1-(2,4-dichlorophenyl)ethanone |
Specifications
4252-78-2 | |
>110°C (230°F) | |
MFCD00000934 | |
957098 | |
VYWPPRLJNVHPEU-UHFFFAOYSA-N | |
2-chloro-1-(2,4-dichlorophenyl)ethanone | |
20250 | |
97% |
52°C to 56°C | |
C8H5Cl3O | |
UN2923 | |
2,2',4'-trichloroacetophenone, 2-chloro-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl chloride, unii-2e35p7hji2, 2-chloro-1-2,4-dichlorophenyl ethan-1-one, ccris 633, ethanone, 2-chloro-1-2,4-dichlorophenyl, 2,2,4-trichloroacetophenone, dsstox_cid_6190, 1-2,4-dichlorophenyl-2-chloroethan-1-one | |
C1=CC(=C(C=C1Cl)Cl)C(=O)CCl | |
223.477 | |
223.49 | |
2,2',4'-Trichloroacetophenone |
Safety and Handling
GHS H Statement
H301-H314-H318-H335
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.