missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R)-(-)-1,2-Diaminocyclohexane, 99%, Thermo Scientific Chemicals

Catalog Number p-3757430
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20439-47-8
C6H14N2
114.19
MFCD00062985,MFCD00063747
SSJXIUAHEKJCMH-UHFFFAOYNA-N
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
43806
(1R,2R)-cyclohexane-1,2-diamine
NC1CCCCC1N
This item is not returnable. View return policy
20439-47-8
C6H14N2
114.19
MFCD00062985,MFCD00063747
SSJXIUAHEKJCMH-UHFFFAOYNA-N
1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
43806
(1R,2R)-cyclohexane-1,2-diamine
NC1CCCCC1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 40.0°C to 45.0°C
Color White to Yellow
Boiling Point 110.0°C to 114.0°C (40.0 Mbar)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H10(NH2)2
Quantity 1 g
Beilstein 13, III, 6
Specific Rotation Condition − 24.50 (26.50°C c=5 1 N HCl 20°)
Specific Rotation − 24.50
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
Formula Weight 114.19
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material (1R, 2R)-(-)-1, 2-Diaminocyclohexane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (1R,2R)-(-)-1,2-Diaminocyclohexane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'