missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 1mL Sterile Serological Pipets
GREENER_CHOICE

Drip-free pipetting design with high optical clarity and maximum accuracy.

Brand:  Greiner Bio-One 604160

 View more versions of this product

Product Code. 10473275

  • £390.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

Shape: Standard
Sterile
1/100
1 mL
Polystyrene
Yes
Individual Plastic Wrap
Serological Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers