missing translation for 'onlineSavingsMsg'
Learn More

12-Crown-4, 97%, Thermo Scientific Chemicals

£48.94 - £286.43

Chemical Identifiers

CAS 294-93-9
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00005103
InChI Key XQQZRZQVBFHBHL-UHFFFAOYSA-N
PubChem CID 9269
ChEBI CHEBI:32399
IUPAC Name 1,4,7,10-tetraoxacyclododecane
SMILES C1COCCOCCOCCO1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10367840
View Documents
Thermo Scientific Acros
187020025
2.5 g Glass bottle
£48.94
2.50g
Please to purchase this item. Need a web account? Register with us today!
10317861
View Documents Promotion Details
Thermo Scientific
187020050
5 g Glass bottle
£80.80
5g
Please to purchase this item. Need a web account? Register with us today!
10741414
View Documents
Thermo Scientific Acros
187020250
25 g Glass bottle
£286.43
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
Specifications

Chemical Identifiers

294-93-9
176.21
XQQZRZQVBFHBHL-UHFFFAOYSA-N
CHEBI:32399
C1COCCOCCOCCO1
C8H16O4
MFCD00005103
9269
1,4,7,10-tetraoxacyclododecane

Specifications

294-93-9
100.0
Colorless to Yellow
238°C
Authentic
C8H16O4
MFCD00005103
1.08
Solubility in water: miscible
C1COCCOCCOCCO1
176.21
CHEBI:32399
97%
12-Crown-4
96.0
16°C
1.0800g/mL
>112°C
96% min. (GC)
1.4600 to 1.463
06,133; 09,126; 12,147; 13,167
12-crown-4, eoct, 12-crown-4 ether, ethylene oxide cyclic tetramer, ccris 157, unii-k6069p2c2a, 12-crown4-ether, 12-crown 4-ether, 5-19-11-00334 beilstein handbook reference, 3,6,9,12-tetraoxacyclododecan
XQQZRZQVBFHBHL-UHFFFAOYSA-N
1,4,7,10-tetraoxacyclododecane
9269
176.21
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if inhaled.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-036-5

Product Identifier
  • 12-Crown-4

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1

Hazard Statement
  • H330-Fatal if inhaled.

Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only