missing translation for 'onlineSavingsMsg'
Learn More
11-(Aminooxy)undecyltriethoxysilane, 95%, Thermo Scientific™ Product Code.: 15476577

11-(Aminooxy)undecyltriethoxysilane, 95%, Thermo Scientific™

Product Code. 15476577
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15476577

Brand: Thermo Scientific Alfa Aesar H33727.MD

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870482-11-4
Molecular Formula C17H39NO4Si
Molecular Weight (g/mol) 349.587
MDL Number MFCD11046089
InChI Key PLHZNMUQNDUOTN-UHFFFAOYSA-N
Synonym o-11-triethoxysilyl undecyl hydroxylamine, 11-aminooxy undecyltriethoxysilane, 11-o-hydroxylamine undecyltriethoxysilane
PubChem CID 45158968
IUPAC Name O-(11-triethoxysilylundecyl)hydroxylamine
SMILES CCO[Si](CCCCCCCCCCCON)(OCC)OCC

Specifications

Boiling Point 145°C to 150°C (0.08mmHg)
Quantity 250 mg
Linear Formula H2NO(CH2)11Si(OCH2CH3)3
Formula Weight 349.59
Percent Purity 95%
Refractive Index 1.4370-1.4410
Sensitivity Moisture sensitive
Chemical Name or Material 11-(Aminooxy)undecyltriethoxysilane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10177498 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
11-(Aminooxy)undecyltriethoxysilane, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.