missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher Chemical 10KG POTASSIUM IODIDE 99.5 % (DRIED)
Brand: Fisher Chemical P/5840/68
2083.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
10KG POTASSIUM IODIDE 99.5 % (DRIED)
This item is not returnable.
View return policy
Description

Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium iodide
Signal Word
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher Chemical 10KG POTASSIUM IODIDE 99.5 % (DRIED) >
Spot an opportunity for improvement?Share a Content Correction