Learn More
Masterflex™ Stopcocks with Luer Connections
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
Brand: Masterflex™ 30600-00
Code : A0
Additional Details : Weight : 0.02520kg
Description
Regulate flow without any leaking using these fittings. Cole-Parmer™ Stopcocks with Luer Connections are available with either a male luer lock or a male luer slip.
- Handle rotates 90°
- PC body and HDPE stem
Specifications
Stopcock | |
Polycarbonate and HDPE | |
Luer |
Polycarbonate, HDPE | |
Male Luer Lock or A Male Luer Slip |