missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid, 97%, Thermo Scientific™

£55.20 - £224.00

Chemical Identifiers

CAS 342404-46-0
Molecular Formula C14H12BNO4S
Molecular Weight (g/mol) 301.12
MDL Number MFCD03086094
InChI Key HXWLCYMHOULBJZ-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-2-indoleboronic acid, 1-phenylsulfonyl-1h-indol-2-yl boronic acid, 1-phenylsulfonyl-1h-indol-2-ylboronic acid, 1-phenylsulfonyl-2-indolylboronic acid, 1-phenylsulfonyl indole-2-boronic acid, 1-benzenesulfonyl indol-2-ylboronic acid, boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl, 1-benzenesulfonyl indol-2-yl boronic acid, acmc-209i5v, 1-phenylsulfonylindole-2-boronic acid
PubChem CID 2776228
IUPAC Name [1-(benzenesulfonyl)indol-2-yl]boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10477992
View Documents
Thermo Scientific Maybridge
CC03312DA
1 g Amber Glass Bottle
£55.20
1g
Please to purchase this item. Need a web account? Register with us today!
10712377
View Documents
Thermo Scientific Maybridge
CC03312DE
5 g Amber Glass Bottle
£224.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

342404-46-0
301.12
HXWLCYMHOULBJZ-UHFFFAOYSA-N
2776228
OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
C14H12BNO4S
MFCD03086094
1-phenylsulfonyl-2-indoleboronic acid, 1-phenylsulfonyl-1h-indol-2-yl boronic acid, 1-phenylsulfonyl-1h-indol-2-ylboronic acid, 1-phenylsulfonyl-2-indolylboronic acid, 1-phenylsulfonyl indole-2-boronic acid, 1-benzenesulfonyl indol-2-ylboronic acid, boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl, 1-benzenesulfonyl indol-2-yl boronic acid, acmc-209i5v, 1-phenylsulfonylindole-2-boronic acid
[1-(benzenesulfonyl)indol-2-yl]boronic acid

Specifications

342404-46-0
C14H12BNO4S
1-phenylsulfonyl-2-indoleboronic acid, 1-phenylsulfonyl-1h-indol-2-yl boronic acid, 1-phenylsulfonyl-1h-indol-2-ylboronic acid, 1-phenylsulfonyl-2-indolylboronic acid, 1-phenylsulfonyl indole-2-boronic acid, 1-benzenesulfonyl indol-2-ylboronic acid, boronic acid,b-1-phenylsulfonyl-1h-indol-2-yl, 1-benzenesulfonyl indol-2-yl boronic acid, acmc-209i5v, 1-phenylsulfonylindole-2-boronic acid
OB(O)C1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1
301.12
301.13
97%
MFCD03086094
HXWLCYMHOULBJZ-UHFFFAOYSA-N
[1-(benzenesulfonyl)indol-2-yl]boronic acid
2776228
1-(Phenylsulfonyl)-1H-indol-2-ylboronic acid
Videos
SDS
Documents

Documents

Product Certifications