missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine dihydrochloride, 98%, Thermo Scientific™

Catalog Number p-7052732
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
34352-59-5
C5H14Cl2N2
173.081
MFCD00012755
AILFRWRYZZVJTL-UHFFFAOYSA-N
1-methylpiperazine dihydrochloride, methylpiperazine, chloride, chloride, piperazine, 1-methyl-dihydrochloride, pubchem15033, methylpiperazine dihydrochloride, n-methylpiperazine dihydrochloride, piperazine, 1-methyl-, dihydrochloride
2734715
1-methylpiperazine;dihydrochloride
CN1CCNCC1.Cl.Cl
This item is not returnable. View return policy
34352-59-5
C5H14Cl2N2
173.081
MFCD00012755
AILFRWRYZZVJTL-UHFFFAOYSA-N
1-methylpiperazine dihydrochloride, methylpiperazine, chloride, chloride, piperazine, 1-methyl-dihydrochloride, pubchem15033, methylpiperazine dihydrochloride, n-methylpiperazine dihydrochloride, piperazine, 1-methyl-, dihydrochloride
2734715
1-methylpiperazine;dihydrochloride
CN1CCNCC1.Cl.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 246°C to 250°C
Color White
Quantity 5 g
Formula Weight 173.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Methylpiperazine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'