Learn More
1-Heptylamine, 98+%, Thermo Scientific Chemicals
£25.93 - £132.00
Chemical Identifiers
CAS | 111-68-2 |
---|---|
Molecular Formula | C7H17N |
Molecular Weight (g/mol) | 115.22 |
MDL Number | MFCD00008244 |
InChI Key | WJYIASZWHGOTOU-UHFFFAOYSA-N |
Synonym | heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 |
PubChem CID | 8127 |
IUPAC Name | heptan-1-amine |
SMILES | CCCCCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429046
|
Thermo Scientific Alfa Aesar
A17922.14 |
25 g |
£25.93
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11408750
|
Thermo Scientific Alfa Aesar
A17922.18 |
50 g |
£37.89
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418750
|
Thermo Scientific Alfa Aesar
A17922.30 |
250 g |
£132.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Heptylamine has been used as an internal standard in the determination of biogenic monoamines and biogenic diamines by the fluorescence-HPLC method. It has also been used to study the effect of hydration on the gaseous structure of protonated heptylamine by infrared photo dissociation (IRPD) spectroscopy and computational chemistry.
Solubility
Soluble in water 6791 mg/L @ 25°C.
Notes
Air sensitive. Store away from oxidizing agents, air, acids.
Chemical Identifiers
111-68-2 | |
115.22 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
8127 | |
CCCCCCCN |
C7H17N | |
MFCD00008244 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
heptan-1-amine |
Specifications
111-68-2 | |
0.775 | |
35°C (95°F) | |
C7H17N | |
1.4245 | |
UN2734 | |
Air Sensitive | |
Soluble in water 6791mg/L @ 25°C. | |
CCCCCCCN | |
115.22 | |
115.22 | |
1-Heptylamine |
-23°C | |
154°C to 156°C | |
Amine-like | |
CH3(CH2)6NH2 | |
MFCD00008244 | |
1731688 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
heptan-1-amine | |
8127 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 203-895-8
missing translation for 'rtecsNumber' : MK0600000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 C
- Skin sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only