missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol, Reagent grade, 98%, Honeywell™

Reagent Grade, 98%

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15611190
View Documents
Honeywell Research Chemicals
126799-250ML
250 mL Poly Bottle N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
15691180
View Documents
Honeywell Research Chemicals
126799-1L
1 L Poly Bottle N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
15601190
View Documents
Honeywell Research Chemicals
126799-2.5L
2.5 L Poly Bottle N/A
N/A
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

112-53-8
186.34
LQZZUXJYWNFBMV-UHFFFAOYSA-N
8193
dodecan-1-ol
C12H26O
MFCD00004753
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
CHEBI:28878
CCCCCCCCCCCCO

Specifications

112-53-8
Colorless
260°C to 262°C
C12H26O
n20/D 1.442(lit.)
UN3077
1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
LQZZUXJYWNFBMV-UHFFFAOYSA-N
dodecan-1-ol
8193
CHEBI:28878
98%
1-Dodecanol
22°C to 26°C
0.833g/mL at 25°C
121°C (249.8°F)
CH3(CH2)11OH
MFCD00004753
1738860
Slightly soluble in water, 1g/L at 23°C
CCCCCCCCCCCCO
186.34
7.4 (vs air)
186.33g/mol
Reagent
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

H319-H410

missing translation for 'dotInformation' : 9/PGIII

missing translation for 'einecsNumber' : 203-982-0

missing translation for 'rtecsNumber' : JR5775000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.