missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)cyclohexanecarbonitrile, 95%, Thermo Scientific™

£31.10 - £127.00

Chemical Identifiers

CAS 112905-95-0
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
InChI Key BWQNUOFYAYIAIY-UHFFFAOYSA-N
Synonym 1-chloromethyl cyclohexanecarbonitrile, 1-chloromethyl cyclohexane-1-carbonitrile, cyclohexanecarbonitrile,1-chloromethyl, acmc-20ds23, 1-chloromethyl-cyclohexanecarbonitrile
PubChem CID 2794920
IUPAC Name 1-(chloromethyl)cyclohexane-1-carbonitrile
SMILES C1CCC(CC1)(CCl)C#N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10137392
SDS View Documents
Thermo Scientific Maybridge
RH01751DA
1 g Amber Glass Bottle
£31.10
1g
Please to purchase this item. Need a web account? Register with us today!
10178204
SDS View Documents
Thermo Scientific Maybridge
RH01751DE
5 g Amber Glass Bottle
£127.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

112905-95-0
157.641
1-chloromethyl cyclohexanecarbonitrile, 1-chloromethyl cyclohexane-1-carbonitrile, cyclohexanecarbonitrile,1-chloromethyl, acmc-20ds23, 1-chloromethyl-cyclohexanecarbonitrile
1-(chloromethyl)cyclohexane-1-carbonitrile
C8H12ClN
BWQNUOFYAYIAIY-UHFFFAOYSA-N
2794920
C1CCC(CC1)(CCl)C#N

Specifications

112905-95-0
C8H12ClN
BWQNUOFYAYIAIY-UHFFFAOYSA-N
1-(chloromethyl)cyclohexane-1-carbonitrile
2794920
95%
97%
1-chloromethyl cyclohexanecarbonitrile, 1-chloromethyl cyclohexane-1-carbonitrile, cyclohexanecarbonitrile,1-chloromethyl, acmc-20ds23, 1-chloromethyl-cyclohexanecarbonitrile
C1CCC(CC1)(CCl)C#N
157.641
157.64
1-(chloromethyl)cyclohexanecarbonitrile
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.