Learn More
1-Chloro-3-iodopropane, 98%, stab. with copper, Thermo Scientific Chemicals
£41.21 - £401.00
Chemical Identifiers
CAS | 6940-76-7 |
---|---|
Molecular Formula | C3H6ClI |
Molecular Weight (g/mol) | 204.435 |
MDL Number | MFCD00001096 |
InChI Key | SFOYQZYQTQDRIY-UHFFFAOYSA-N |
Synonym | propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane |
PubChem CID | 81363 |
IUPAC Name | 1-chloro-3-iodopropane |
SMILES | C(CCl)CI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11465573
|
Thermo Scientific Alfa Aesar
A13489.14 |
25 g |
£41.21
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475573
|
Thermo Scientific Alfa Aesar
A13489.22 |
100 g |
£116.58
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485573
|
Thermo Scientific Alfa Aesar
A13489.36 |
500 g |
£401.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.
Solubility
Difficult to mix.
Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
6940-76-7 | |
204.435 | |
SFOYQZYQTQDRIY-UHFFFAOYSA-N | |
81363 | |
C(CCl)CI |
C3H6ClI | |
MFCD00001096 | |
propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane | |
1-chloro-3-iodopropane |
Specifications
6940-76-7 | |
170°C to 172°C | |
C3H6ClI | |
MFCD00001096 | |
Light sensitive | |
Difficult to mix. | |
C(CCl)CI | |
204.435 | |
204.44 | |
1-Chloro-3-iodopropane, stabilized with copper |
1.904 | |
>110°C (230°F) | |
1.5463 | |
1731115 | |
propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane | |
SFOYQZYQTQDRIY-UHFFFAOYSA-N | |
1-chloro-3-iodopropane | |
81363 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 230-088-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.