missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

£34.82 - £323.61

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10485851
SDS View Documents Promotion Details
Thermo Scientific Acros
423495000
500 mL EcoSafPak™ Glass Bottle
£34.82
500mL
Please to purchase this item. Need a web account? Register with us today!
10321311
SDS View Documents Promotion Details
Thermo Scientific Acros
423490010
1 L EcoSafPak™ Glass Bottle
£52.57
1 L
Please to purchase this item. Need a web account? Register with us today!
10115753
SDS View Documents
Thermo Scientific Acros
423490025
2.5 L Glass Bottle
£117.06
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11994591
SDS View Documents
Thermo Scientific Acros
423490100
10 L Plastic Drum
£323.61
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur., Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.