Learn More
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
£24.52 - £244.22
Chemical Identifiers
CAS | 112-71-0 |
---|---|
Molecular Formula | C14H29Br |
Molecular Weight (g/mol) | 277.29 |
MDL Number | MFCD00000228 |
InChI Key | KOFZTCSTGIWCQG-UHFFFAOYSA-N |
Synonym | myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo |
PubChem CID | 8208 |
IUPAC Name | 1-bromotetradecane |
SMILES | CCCCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483087
|
Thermo Scientific Alfa Aesar
A16082.22 |
100 g |
£24.52
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493087
|
Thermo Scientific Alfa Aesar
A16082.36 |
500 g |
£60.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403097
|
Thermo Scientific Alfa Aesar
A16082.0E |
2500 g |
£244.22
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
112-71-0 | |
277.29 | |
KOFZTCSTGIWCQG-UHFFFAOYSA-N | |
8208 | |
CCCCCCCCCCCCCCBr |
C14H29Br | |
MFCD00000228 | |
myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo | |
1-bromotetradecane |
Specifications
112-71-0 | |
1.018 | |
158°C (316°F) | |
1.46 | |
1742640 | |
Not miscible in water. | |
CCCCCCCCCCCCCCBr | |
277.29 | |
277.3 | |
1-Bromotetradecane |
4°C to 6°C | |
175°C to 178°C (20 mmHg) | |
C14H29Br | |
MFCD00000228 | |
myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo | |
KOFZTCSTGIWCQG-UHFFFAOYSA-N | |
1-bromotetradecane | |
8208 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 203-999-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only