missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopropane, 99%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 106-94-5
Molecular Formula C3H7Br
Molecular Weight (g/mol) 122.993
MDL Number MFCD00000254
InChI Key CYNYIHKIEHGYOZ-UHFFFAOYSA-N
Synonym n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
PubChem CID 7840
ChEBI CHEBI:47105
IUPAC Name 1-bromopropane
SMILES CCCBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11448693
SDS View Documents
Thermo Scientific Alfa Aesar
A10461.AK
250 mL N/A
N/A
11438693
SDS View Documents
Thermo Scientific Alfa Aesar
A10461.AU
1000 mL N/A
N/A
11458693
SDS View Documents
Thermo Scientific Alfa Aesar
A10461.0J
5000 mL N/A
N/A
Description

Description

1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

Solubility
Miscible with ethanol, water and diethyl ether.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

106-94-5
122.993
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
7840
1-bromopropane
C3H7Br
MFCD00000254
n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
CHEBI:47105
CCCBr

Specifications

106-94-5
1.353
21°C (70°F)
CH3CH2CH2Br
MFCD00000254
505936
14,7845
Miscible with ethanol,water and diethyl ether.
CCCBr
122.993
CHEBI:47105
99%
-110°C
70°C to 71°C
C3H7Br
1.434
UN2344
Light sensitive
n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide
CYNYIHKIEHGYOZ-UHFFFAOYSA-N
1-bromopropane
7840
123
1-Bromopropane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H360-H373-H315-H319-H335-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335-H336-H350-H360FD-H373

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BROMOPROPANES

missing translation for 'einecsNumber' : 203-445-0

missing translation for 'rtecsNumber' : TX4110000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.