Learn More
1-Bromopinacolone, 93%, Thermo Scientific Chemicals
£30.80 - £114.00
Chemical Identifiers
CAS | 5469-26-1 |
---|---|
Molecular Formula | C6H11BrO |
Molecular Weight (g/mol) | 179.06 |
MDL Number | MFCD00000206 |
InChI Key | SAIRZMWXVJEBMO-UHFFFAOYSA-N |
Synonym | 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german |
PubChem CID | 21642 |
IUPAC Name | 1-bromo-3,3-dimethylbutan-2-one |
SMILES | CC(C)(C)C(=O)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10267180
|
Thermo Scientific Acros
306410050 |
5 mL | Glass bottle |
£30.80
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10331094
|
Thermo Scientific Acros
306410250 |
25 mL | Glass bottle |
£114.00
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5469-26-1 | |
179.06 | |
SAIRZMWXVJEBMO-UHFFFAOYSA-N | |
21642 | |
CC(C)(C)C(=O)CBr |
C6H11BrO | |
MFCD00000206 | |
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german | |
1-bromo-3,3-dimethylbutan-2-one |
Specifications
5469-26-1 | |
100.0 | |
Brown-Yellow to Yellow | |
189°C to 192°C | |
Authentic | |
C6H11BrO | |
1.4656 to 1.4676 | |
1.33 | |
SAIRZMWXVJEBMO-UHFFFAOYSA-N | |
1-bromo-3,3-dimethylbutan-2-one | |
21642 | |
93% | |
1-Bromopinacolone |
91.0 | |
-10°C | |
1.3300g/mL | |
75°C | |
91% min. (GC) | |
(CH3)3CCOCH2Br | |
MFCD00000206 | |
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german | |
CC(C)(C)C(=O)CBr | |
179.06 | |
179.06 | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-794-
RUO – Research Use Only