Learn More
1'-Acetonaphthone, 95%, Thermo Scientific Chemicals
£63.80 - £227.00
Chemical Identifiers
CAS | 941-98-0 |
---|---|
Molecular Formula | C12H10O |
Molecular Weight (g/mol) | 170.21 |
MDL Number | MFCD00004013 |
InChI Key | QQLIGMASAVJVON-UHFFFAOYSA-N |
Synonym | 1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone |
PubChem CID | 13663 |
IUPAC Name | 1-naphthalen-1-ylethanone |
SMILES | CC(=O)C1=CC=CC2=CC=CC=C21 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10386831
|
Thermo Scientific Acros
400092500 |
250 g |
£63.80
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10215041
|
Thermo Scientific Acros
400090010 |
1 kg |
£227.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
941-98-0 | |
170.21 | |
QQLIGMASAVJVON-UHFFFAOYSA-N | |
13663 | |
CC(=O)C1=CC=CC2=CC=CC=C21 |
C12H10O | |
MFCD00004013 | |
1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone | |
1-naphthalen-1-ylethanone |
Specifications
941-98-0 | |
100.0 | |
Yellow | |
302.0°C | |
Authentic | |
Glass bottle | |
C10H7COCH3 | |
MFCD00004013 | |
1'-acetonaphthone, 1-acetonaphthone, 1-acetylnaphthalene, methyl 1-naphthyl ketone, 1-acetonaphthalene, 1-1-naphthalenyl ethanone, ethanone, 1-1-naphthalenyl, 1-naphthalen-1-yl ethanone, 1-naphthyl methyl ketone, alpha-acetonaphthone | |
QQLIGMASAVJVON-UHFFFAOYSA-N | |
1-naphthalen-1-ylethanone | |
13663 | |
95% | |
1'-Acetonaphthone, 95% |
94.0 | |
12.0°C | |
1.1190g/mL | |
>110°C | |
94% min. (GC) | |
C12H10O | |
1.6270 to 1.6290 | |
1.119 | |
Solubility in water: immiscible | |
CC(=O)C1=CC=CC2=CC=CC=C21 | |
170.21 | |
170.21 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 213-384-1
missing translation for 'rtecsNumber' : AL2982800
missing translation for 'tsca' : TSCA
- 1'-Acetonaphthone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only