missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane, 98+%, Thermo Scientific Chemicals

Product Code. 17426742
Click to view available options
Quantity:
100 g
500 g
2 kg
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Diaminohexane is mainly used as a monomer to make nylon 6-6. Its derivative hexamethylene diisocyanate (HDI) is used in the production of polyurethane. It acts as a cross-linking agent in epoxy resins. Other applications include coatings, lubricants and water treatment products.

Solubility
Soluble in water. Slightly soluble in ether, alcohol and benzene.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, strong oxidizing agents, acid chlorides, carbon dioxide and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6H16N2
MDL Number MFCD00008243
InChI Key NAQMVNRVTILPCV-UHFFFAOYSA-N
Synonym 1,6-hexanediamine, 1,6-diaminohexane, hexamethylenediamine, hmda, 1,6-hexylenediamine, 1,6-hexamethylenediamine, 1,6-diamino-n-hexane, hexamethylene diamine, hexylenediamine, 1,6-hexanediamine solution
PubChem CID 16402
ChEBI CHEBI:39618
IUPAC Name hexane-1,6-diamine

Specifications

Melting Point 39°C to 43°C
Density 0.84
pH 12.4
Boiling Point 199°C to 204°C
Flash Point 85°C
Odor Amine-like
Linear Formula H2N(CH2)6NH2
Quantity 2 kg
UN Number UN2280
Beilstein 1098307
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,4695
Solubility Information Soluble aq. soln.. Slightly soluble in ether,alcohol and benzene.
SMILES NCCCCCCN
Molecular Weight (g/mol) 116.21
Formula Weight 116.21
Percent Purity ≥98%
Chemical Name or Material 1,6-Diaminohexane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.