missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diazabicyclo[4.3.0]non-5-ene, 98%, Thermo Scientific Chemicals

£16.38 - £136.68

Chemical Identifiers

CAS 3001-72-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00005554
InChI Key SGUVLZREKBPKCE-UHFFFAOYSA-N
Synonym 1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene
PubChem CID 76349
IUPAC Name 2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine
SMILES C1CN2CCCN=C2C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11465483
View Documents
Thermo Scientific Alfa Aesar
A13437.06
5 g
£16.38
5g
Please to purchase this item. Need a web account? Register with us today!
11475483
View Documents
Thermo Scientific Alfa Aesar
A13437.14
25 g
£43.42
25g
Please to purchase this item. Need a web account? Register with us today!
11485483
View Documents
Thermo Scientific Alfa Aesar
A13437.22
100 g
£136.68
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.

Solubility
Miscible with water.

Notes
Air, light and moisture sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
Specifications

Chemical Identifiers

3001-72-7
124.19
SGUVLZREKBPKCE-UHFFFAOYSA-N
76349
C1CN2CCCN=C2C1
C7H12N2
MFCD00005554
1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene
2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine

Specifications

3001-72-7
94°C (201°F)
MFCD00005554
2417
SGUVLZREKBPKCE-UHFFFAOYSA-N
2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine
76349
98%
95°C to 98°C (7 mmHg)
C7H12N2
UN3267
1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene
C1CN2CCCN=C2C1
124.19
124.19
1,5-Diazabicyclo[4.3.0]non-5-ene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only