Learn More
1-(4-Fluorophenyl)piperazine dihydrochloride, 98%, Thermo Scientific™
£29.32 - £309.49
Chemical Identifiers
CAS | 64090-19-3 |
---|---|
Molecular Formula | C10H15Cl2FN2 |
Molecular Weight (g/mol) | 253.142 |
MDL Number | MFCD00012765 |
InChI Key | DZQVAQAZQDURKX-UHFFFAOYSA-N |
Synonym | 1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p |
PubChem CID | 16211916 |
IUPAC Name | 1-(4-fluorophenyl)piperazine;dihydrochloride |
SMILES | C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
64090-19-3 | |
253.142 | |
DZQVAQAZQDURKX-UHFFFAOYSA-N | |
16211916 | |
C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl |
C10H15Cl2FN2 | |
MFCD00012765 | |
1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p | |
1-(4-fluorophenyl)piperazine;dihydrochloride |
Specifications
64090-19-3 | |
C10H15Cl2FN2 | |
UN2811 | |
1-4-fluorophenyl piperazine dihydrochloride, 1-4-fluorophenyl piperazinedihydrochloride, 1-4-fluorophenyl piperazine 2hcl, piperazine, 1-4-fluorophenyl-, dihydrochloride, 1-4-fluorophenyl piperazine;dihydrochloride, para-fluorophenylpiperazine dihydrochloride, zlchem 337, pubchem15283, acmc-209nk7, ksc352o8p | |
C1CN(CCN1)C2=CC=C(C=C2)F.Cl.Cl | |
253.142 | |
253.15 | |
1-(4-Fluorophenyl)piperazine dihydrochloride |
∽234°C (decomposition) | |
MFCD00012765 | |
4826840 | |
DZQVAQAZQDURKX-UHFFFAOYSA-N | |
1-(4-fluorophenyl)piperazine;dihydrochloride | |
16211916 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures