Learn More
1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals
£153.00 - £608.00
Chemical Identifiers
CAS | 2425-79-8 |
---|---|
Molecular Formula | C10H18O4 |
Molecular Weight (g/mol) | 202.25 |
MDL Number | MFCD00005146 |
InChI Key | SHKUUQIDMUMQQK-UHFFFAOYNA-N |
Synonym | 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane |
PubChem CID | 17046 |
IUPAC Name | 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane |
SMILES | C(CCOCC1CO1)COCC1CO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10235820
|
Thermo Scientific Acros
154315000 |
500 g |
£153.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10105693
|
Thermo Scientific Acros
154310025 |
2.5 kg |
£608.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2425-79-8 | |
202.25 | |
SHKUUQIDMUMQQK-UHFFFAOYNA-N | |
17046 | |
C(CCOCC1CO1)COCC1CO1 |
C10H18O4 | |
MFCD00005146 | |
1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane | |
2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane |
Specifications
2425-79-8 | |
Yellow | |
266.0°C | |
Authentic | |
Glass bottle | |
1.4600 to 1.4650 | |
1.1 | |
Solubility in water: 55,6g/L (20°C). | |
C(CCOCC1CO1)COCC1CO1 | |
202.25 | |
<15 mPa.s (25°C) | |
60% | |
Liquid |
60 | |
1.1000g/mL | |
110°C | |
55% min. (GC) | |
C10H18O4 | |
MFCD00005146 | |
1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane | |
SHKUUQIDMUMQQK-UHFFFAOYNA-N | |
2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane | |
17046 | |
202.25 | |
Technical | |
1, 4-Butanediol diglycidyl ether, 60% |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful to aquatic life with long l
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 219-371-7
missing translation for 'rtecsNumber' : EJ5100000
missing translation for 'tsca' : TSCA
Recommended Storage : This product unavoidably and progressively degrades on storage
- 1,4-Butanediol diglycidyl ether
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only