Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
£31.85 - £53.55
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Overall Length (Metric) | Price | Quantity & Availability | ||||
15208753
|
Trajan Scientific and Medical™
0624245 |
0.025 mm | 250 mm |
£40.50
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15218753
|
Trajan Scientific and Medical™
0624249 |
0.025 mm | 500 mm |
£53.55
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15208763
|
Trajan Scientific and Medical™
0624265 |
0.05 mm | 250 mm |
£40.50
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15228773
|
Trajan Scientific and Medical™
0624312 |
0.1 mm | 100 mm |
£33.90
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15258773
|
Trajan Scientific and Medical™
0624319 |
0.1 mm | 500 mm |
£53.55
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15248773
|
Trajan Scientific and Medical™
0624315 |
0.1 mm | 250 mm |
£40.50
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
12726248
|
Trajan Scientific and Medical™
0624279 |
0.075 mm | 500 mm |
£53.55
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
15288773
|
Trajan Scientific and Medical™
0624349 |
0.15 mm | 500 mm |
£53.55
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
12706248
|
Trajan Scientific and Medical™
0624273 |
0.075 mm | 150 mm |
£36.15
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
12716248
|
Trajan Scientific and Medical™
0624275 |
0.075 mm | 250 mm |
£40.50
Pack of 2 |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
15218773
|
Trajan Scientific and Medical™
0624311 |
0.1 mm | 50 mm |
£31.85
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15238773
|
Trajan Scientific and Medical™
0624313 |
0.1 mm | 150 mm |
£36.15
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15268773
|
Trajan Scientific and Medical™
0624342 |
0.15 mm | 100 mm |
£33.90
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15278743
|
Trajan Scientific and Medical™
0624241 |
0.025 mm | 50 mm |
£31.85
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15278773
|
Trajan Scientific and Medical™
0624345 |
0.15 mm | 250 mm |
£40.50
Pack of 2 |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
15288743
|
Trajan Scientific and Medical™
0624242 |
0.025 mm | 100 mm |
£33.90
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
15298743
|
Trajan Scientific and Medical™
0624243 |
0.025 mm | 150 mm |
£36.15
Pack of 2 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
Description
- Fused silica internal wall provides smooth surface, free of imperfections
- PEEKsil compatible with most organic solvents and resistant to strong acids
- Effective pH range of 0 to 10
- Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Tubing | |
2/Pk. |