missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis[4-(4-pyridyl)-2-thiazolyl]benzene, 97%, Thermo Scientific™

Product Code. 15464898
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The derivatives of this compound has been synthesized and then reacted with nitrate salts of copper, cadmium, and cobalt and copper hexafluororacetylacetonate to produce new one-dimensional and two-dimensional coordination polymers which display a variety of structural motifs.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421261-85-9
Molecular Formula C22H14N4S2
Molecular Weight (g/mol) 398.502
MDL Number MFCD22682999
InChI Key BAIVNAOKZPUEIV-UHFFFAOYSA-N
Synonym 1,3-bis 4-4-pyridyl-2-thiazolyl benzene, 4-2-3-4-pyridin-4-yl-1,3-thiazol-2-yl phenyl-1,3-thiazol-4-yl pyridine
PubChem CID 86277494
IUPAC Name 4-pyridin-4-yl-2-[3-(4-pyridin-4-yl-1,3-thiazol-2-yl)phenyl]-1,3-thiazole
SMILES C1=CC(=CC(=C1)C2=NC(=CS2)C3=CC=NC=C3)C4=NC(=CS4)C5=CC=NC=C5

Specifications

Melting Point 290°C to 296°C
Quantity 5 g
Formula Weight 398.49
Percent Purity 97%
Chemical Name or Material 1,3-Bis[4-(4-pyridyl)-2-thiazolyl]benzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'